Orders LinkContactUsLinkComponent
Multi-parameter meter, handheld, Professional Plus
Multi-parameter meter, handheld, Professional Plus
Quatro cable/bulkhead (includes conductivity/temperature sensor), 20 m
Catalogus # 665-0205
Leverancier:   YSI ENVIRONMENTAL
Multi-parameter meter, handheld, Professional Plus
Catalogus # 665-0205
Leverancier:  YSI ENVIRONMENTAL
Multi-parameter meter, handheld, Professional Plus
Multi-parameter meter, handheld, Professional Plus
Quatro cable/bulkhead (includes conductivity/temperature sensor), 20 m
Price
3 570,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The Professional Plus is one of the most versatile multi-parameter handheld meters on the market. A combination of up to 11 parameters can be measured on a single bulkhead, including pH, ORP, ammonia, nitrate, chloride, DO, conductivity, salinity, TDS, barometric pressure and temperature, meaning only one cable is required.
  • Rubber, over-moulded IP 67 housing (even without battery cover)
  • Rugged cable sensors with military specification, waterproof connectors
  • USB connection and Data Manager software with GLP functions
  • 5000 data sets memory
  • Easy to read graphic, backlit display and keypad, and multiple language interface: English, German, French, Spanish, Italian, Portuguese and Norwegian
The Quatro cable/bulkhead allows the measuring of up to six parameters at one time (e.g. conductivity, temperature, dissolved oxygen, 2 mV readings and barometric pressure) and has a combined weight of 525 g.
The sensors are totally interchangeable and easily replaceable by the user, and there are a variety of cable lengths available (1, 4, 10, 20 and 30 m). The Pro Plus can be used with other bulkheads to measure one, two or three parameters at the same time.
Three year instrument warranty, two year cable warranty.
Bestelinformatie: The instrument Professional Plus is supplied with Data Manager software, USB cable and USB saddle. Quatro cable and sensors need to be ordered separately.
Specificaties
  • Description:
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 20 m
  • Seq:
    04
  • Cat. No.:
    665-0205
Accessories
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 664-0025
    Soft-sided carrying case
    Log in voor beschikbaarheid
    1 * 1 ST - 286,00 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 664-0024
    Hard-sided carrying case
    Log in voor beschikbaarheid
    1 * 1 ST - 660,00 €
Productinformatie
  • Catalogus #
    Description
    Pk
    Beschikbaarheid
    Price
  • YSIA005580
    Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each)
    1 KIT
    Log in voor beschikbaarheid
    1 * 1 KIT - 229,00 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 665-0069
    Small tripod for flat surface
    Log in voor beschikbaarheid
    1 * 1 ST - 70,10 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 664-0026
    Cable weight, 140 g, attached to stainless steel probe guard
    Log in voor beschikbaarheid
    1 * 1 ST - 61,50 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 665-0235
    Flow cell for Quatro cable/bulkhead
    Log in voor beschikbaarheid
    1 * 1 ST - 880,00 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 665-0091
    pH/ORP kit (must be screwed into probe)
    Log in voor beschikbaarheid
    1 * 1 ST - 842,00 €
Productinformatie
  • Catalogus #
    Description
    Beschikbaarheid
    Price
  • 665-0090
    pH kit (must be screwed into probe)
    Log in voor beschikbaarheid
    1 * 1 ST - 624,00 €