Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, dNTPs

VWR®, dNTPs

dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 16×250 μl, 1 SET
Catalog # 733-1855
Supplier:   VWR Chemicals
VWR®, dNTPs
Catalog # 733-1855
Supplier:  VWR Chemicals
CAS Number:  
VWR®, dNTPs
VWR®, dNTPs

dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 16×250 μl, 1 SET

CAS Number:  
Price
625,00 €
Connexion Pour la disponibilité
1 * 1 SET
Product Details & Documents

Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. dNTP solutions are also available in sets of four individual dNTPs, each 100 mm. Both are convenient for use in DNA polymerisation reactions, DNA labelling and sequencing processes.

  • Available as pre-mixed 10 or 25 mM solutions, or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7,3 to 7,5
Spécifications

Spécifications

  • Pk:
    1 SET
  • Description:
    dNTP set, separate vials of dA, dC, dG, dT, each 100 mM, 16×250 μl

Spécifications