ContactUsLinkComponent
BioSpectrum 800 2D Imaging System. With MegaCam camera: 16 bit monochrome, digital camera and mount. 3326 x 2504 resolution with bin ning capability (8.3 MP). Peltier cooling to -50C from ambient. Camera mount and 24-70mm, f/2.8 lens (manual zoom len 1 * 1 items
Catalog # UVPA97-0500-02
BioSpectrum 800 2D Imaging System. With MegaCam camera: 16 bit monochrome, digital camera and mount. 3326 x 2504 resolution with bin ning capability (8.3 MP). Peltier cooling to -50C from ambient. Camera mount and 24-70mm, f/2.8 lens (manual zoom len 1 * 1 items
Catalog # UVPA97-0500-02
Frequently Bought Together