ContactUsLinkComponent
BioSpectrum 615, MOTORISED fixed 50mm, f/1.2, MOTORISED Lift: 615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digita l camera and mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and 50mm, f 1 * 1 items
Catalog # UVPA97-0400-02
BioSpectrum 615, MOTORISED fixed 50mm, f/1.2, MOTORISED Lift: 615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digita l camera and mount, with binning capability (3.2 Mp). Peltier cooling to -60C from ambient. Camera mount and 50mm, f 1 * 1 items
Catalog # UVPA97-0400-02
Frequently Bought Together