Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pipette tips, universal

Standard, 50 - 1000 µl, 70,5 mm, Blue, Bulk
  BOET03.023.0101
Pipette tips, universal
  BOET03.023.0101
Pipette tips, universal
Pipette tips, universal

Standard, 50 - 1000 µl, 70,5 mm, Blue, Bulk

 
18,60 €
   
1 * 1.000 ST
 

These tips are made from selected, hydrophobic PP.

  • Optimal surface quality
  • Precise and centric opening of tip
  • For optimal use on single and multi-channel pipettes
  • Autoclavable at 121 °C/2,1 bar
  • High accuracy
 
  • Packed:
    Bulk
  • Volume:
    50 - 1000 µl
  • Length:
    70,5 mm
  • Seq:
    03
  • Tip style:
    Standard
  • Tip colour:
    Blue