Microscope slides
Microscope slides with one end frosted on both sides, 1,0 - 1,2 mm, 76×26 mm
Catalog # SMITMC2952/TF76X26
Supplier: SMITH SCIENTIFIC
Jump to:
Product Details & Documents
These slides are made of glass with ground edges.
Specifications
- Description:Microscope slides with one end frosted on both sides
- L×W:76×26 mm
- Seq:004
- Thickness:1,0 - 1,2 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



