Blades
Medical blades, single edge, Gem® Scientific, 3-Facet, degreased steel back, 10 blades per dispenser, Stainless steel, 0,22 mm, 38,1×19 mm
Catalog # PERSAGBL-7039-0000
Supplier: PERSONNA MEDICAL
Jump to:
Product Details & Documents
Single-use blades for a variety of applications.
Specifications
- Description:Medical blades, single edge, Gem® Scientific, 3-Facet, degreased steel back, 10 blades per dispenser
- L×W:38,1×19 mm
- Material:Stainless steel
- Thickness:0,22 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



