Universal absorbent rolls
Heavyweight universal roll, 1 roll/pack, 45 m×400 mm
MSGA9950
: asecos
Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Absorbency of a capacity up to 16 times of its own weight
- High rib and rub resistance
- Standard material for small leaks and prevention
- Heavyweight material for small leaks or the absortion of spillage
- Rolls are perforated to take sufficient material as required
- Description:Heavyweight universal roll
- L×W:45 m×400 mm
- Seq:03
- Packed:1 roll/pack
Accessories
- Description
- Description
- Description
- MSGA9905Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm width1 * 1 ST - 69,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



