Universal absorbent rolls
Heavyweight universal roll, 2 rolls/pack, 45 m×800 mm
Catalog # MSGA9949
Supplier: asecos
Jump to:
Product Details & Documents
Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
- Absorbency of a capacity up to 16 times of its own weight
- High rib and rub resistance
- Standard material for small leaks and prevention
- Heavyweight material for small leaks or the absortion of spillage
- Rolls are perforated to take sufficient material as required
Specifications
- Description:Heavyweight universal roll
- L×W:45 m×800 mm
- Seq:04
- Packed:2 rolls/pack
Accessories
Product Information
- Catalog #AvailabilityPrice
- Roll stand, wide with cutting edge for rolls up to 80 cm widthRoll stand, wide with cutting edge for rolls up to 80 cm widthMSGA9909Log in for availability1 * 1 ST - 244,00 €
Product Information
- Catalog #AvailabilityPrice
- Roll stand, small, with cutting edge for rolls up to 40 cm widthRoll stand, small, with cutting edge for rolls up to 40 cm widthMSGA9908Log in for availability1 * 1 ST - 64,90 €
Product Information
- Catalog #AvailabilityPrice
- Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm widthRack with cutting edge for rolls which need to be ready at hand, up to 40 cm widthMSGA9905Log in for availability1 * 1 ST - 69,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



