Orders LinkContactUsLinkComponent
Universal absorbent pillows
Universal absorbent pillows
Universal pillow, 10 pillows/pack, 460×460 mm
Catalog # MSGA9943
Supplier:   asecos
Universal absorbent pillows
Catalog # MSGA9943
Supplier:  asecos
Universal absorbent pillows
Universal absorbent pillows
Universal pillow, 10 pillows/pack, 460×460 mm
Price
137,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Universal pillows for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for those hard to reach places
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
Specifications
  • Description:
    Universal pillow
  • L×W:
    460×460 mm
  • Packed:
    10 pillows/pack