Orders LinkContactUsLinkComponent
Absorbent pillows
Absorbent pillow, 10 pillows/pack, 460×460 mm
Catalog # MSGA9916
Supplier:   asecos
Absorbent pillows
Catalog # MSGA9916
Supplier:  asecos
Absorbent pillows
Absorbent pillows
Absorbent pillow, 10 pillows/pack, 460×460 mm
Price
79,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Special pillows for safe absorption of acids, alkalis, aggressive chemicals and oil.
Specifications
  • Description:
    Absorbent pillow
  • L×W:
    460×460 mm
  • Packed:
    10 pillows/pack