Orders LinkContactUsLinkComponent
Absorbent pillows
Absorbent pillows
Absorbent pillow, 20 pillows/pack, 250×250 mm
Catalog # MSGA9910
Supplier:   asecos
Absorbent pillows
Catalog # MSGA9910
Supplier:  asecos
Absorbent pillows
Absorbent pillows
Absorbent pillow, 20 pillows/pack, 250×250 mm
Price
141,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Special pillows for safe absorption of acids, alkalis, aggressive chemicals and oil.
Specifications
  • Description:
    Absorbent pillow
  • L×W:
    250×250 mm
  • Packed:
    20 pillows/pack