Orders LinkContactUsLinkComponent
Universal socks
Universal socks
Universal sorbent snakes, 6 per pack, 75×3000 mm
Catalog # MSGA11347
Supplier:   asecos
Universal socks
Catalog # MSGA11347
Supplier:  asecos
Universal socks
Universal socks
Universal sorbent snakes, 6 per pack, 75×3000 mm
Price
106,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Universal socks for safe absorption of oil, water, cooling liquids and non aggressive chemicals.
  • Optimal for long term use
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
For containment and absorption of larger leaks.
Specifications
  • Description:
    Universal sorbent snakes
  • Packed:
    6 per pack
  • Ø×L:
    75×3000 mm