Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Universal absorbent rolls

Standard universal roll, 1 roll/pack, 90 m×800 mm
  MSGA11317
 :   asecos
Universal absorbent rolls
  MSGA11317
 :  asecos
Universal absorbent rolls
Universal absorbent rolls

Standard universal roll, 1 roll/pack, 90 m×800 mm

 
246,00 €
   
1 * 1 ST
 

Universal rolls for safe absorption of oil, water, cooling liquids and non aggressive chemicals.

  • Absorbency of a capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Standard material for small leaks and prevention
  • Heavyweight material for small leaks or the absortion of spillage
  • Rolls are perforated to take sufficient material as required
 
  • Description:
    Standard universal roll
  • L×W:
    90 m×800 mm
  • Seq:
    02
  • Packed:
    1 roll/pack
Accessories

 

  •  
    Description
     
     
  • MSGA9909
    Roll stand, wide with cutting edge for rolls up to 80 cm width
       
    1 * 1 ST - 244,00 €

 

  •  
    Description
     
     
  • MSGA9908
    Roll stand, small, with cutting edge for rolls up to 40 cm width
       
    1 * 1 ST - 64,90 €

 

  •  
    Description
     
     
  • MSGA9905
    Rack with cutting edge for rolls which need to be ready at hand, up to 40 cm width
       
    1 * 1 ST - 69,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.