Orders LinkContactUsLinkComponent
Closures
Closures
Closure, thrust and turn-degasser with and Gore Duravent D15 valve, 28 mm, Red/clear, PP
Catalog # RIXI8-2206-28-R
Closures
Catalog # RIXI8-2206-28-R
Closures
Closures
Closure, thrust and turn-degasser with and Gore Duravent D15 valve, 28 mm, Red/clear, PP
Price
48,20 €
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Thread:
    28 mm
  • Colour:
    Red/clear
  • Description:
    Closure, thrust and turn-degasser with and Gore Duravent D15 valve
  • Material:
    PP
  • Seq:
    4