Closures for test tubes and flasks
Identi-plug foam plugs, Ø 14 - 20 mm
JAECL800-B
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
- Description:Identi-plug foam plugs, Ø 14 - 20 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



