Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

PIG® Poly Drip Decks

Drip deck, 167 L sump capacity, 2722 kg load capacity, White/black, 31,8 kg, 1320×1320×150 mm
  121-0187
 :   NEW PIG
PIG® Poly Drip Decks
  121-0187
 :  NEW PIG
PIG® Poly Drip Decks
PIG® Poly Drip Decks

Drip deck, 167 L sump capacity, 2722 kg load capacity, White/black, 31,8 kg, 1320×1320×150 mm

 
391,00 €
   
1 * 1 ST
 

These poly drip decks capture drips from containers and drums before they cause slip-and-fall hazards. These decks keep leaky drums and containers from creating puddles and slick spots on the floors.

  • Low-profile 15 cm high design is easy to load or unload; pumps and funnels are easy to access
  • Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
  • Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Translucent white sidewalls allow easy visual inspection

Partly made from post-industrial recycled polyethtylene.

 
  • Weight:
    31,8 kg
  • Colour:
    White/black
  • Description:
    Drip deck, 167 L sump capacity, 2722 kg load capacity
  • Seq:
    03
  • W×D×H:
    1320×1320×150 mm