Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

60 - 75, 0,5, 300 mm
  ALLA0150FG075/15QP
Baumé hydrometers
  ALLA0150FG075/15QP
Baumé hydrometers
Baumé hydrometers

60 - 75, 0,5, 300 mm

 
52,30 €
   
1 * 1 ST
 

Baumé hydrometers calibrated at 15 °C.

 
  • Length:
    300 mm
  • Seq:
    28
  • Division:
    0,5
  • Range:
    60 - 75