Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

5 - 15, 0,1, 300 mm
  ALLA0050FG015/15QP
Baumé hydrometers
  ALLA0050FG015/15QP
Baumé hydrometers
Baumé hydrometers

5 - 15, 0,1, 300 mm

 
50,80 €
   
1 * 1 ST
 

Baumé hydrometers calibrated at 15 °C.

 
  • Length:
    300 mm
  • Seq:
    06
  • Division:
    0,1
  • Range:
    5 - 15