Pasteur pipettes, Corning®
Non plugged, borosilicate glass, 6,5 mm, 146 mm, -, 200/pack
Supplier: Corning
Jump to:
Product Details & Documents
Soda-lime or borosilicate glass.
- Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
- Features a constriction one inch below the top to facilitate plugging
- Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.
Specifications
- Ext.Ø:6,5 mm
- Packed:200/pack
- Sterile:-
- Description:Non plugged, borosilicate glass
- Length:146 mm
- Seq:02
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



