Pasteur pipettes, Corning®
Non plugged, long tip, soda-lime glass, 6,5 mm, 330 mm, -, 100/case
Catalog # 612-5198
Supplier: Corning
Jump to:
Product Details & Documents
Soda-lime or borosilicate glass.
- Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
- Features a constriction one inch below the top to facilitate plugging
- Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, haematology and serology studies, as well as blood bank and clinical chemistry procedures.
Documents
Specifications
- Ext.Ø:6,5 mm
- Packed:100/case
- Sterile:-
- Description:Non plugged, long tip, soda-lime glass
- Length:330 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



