Orders LinkContactUsLinkComponent
Cell culture chambers, Corning® CellSTACK®
5 chambers, 625 - 950 ml, 3180 cm²
  734-4061
   Low Manufacturing Impact
 :   Corning
Cell culture chambers, Corning® CellSTACK®
  734-4061
 :  Corning
   Low Manufacturing Impact
Cell culture chambers, Corning® CellSTACK®
Cell culture chambers, Corning® CellSTACK®
5 chambers, 625 - 950 ml, 3180 cm²
1 770,00 €
1 * 8 ST
PS, sterile. The Corning® CellSTACK® culture chambers are available in five sizes and with three different surface treatments.
  • Two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
  • Standard 33 mm threaded caps have 0,2 μm pore non-wettable membranes sealed directly to the caps to allow gas exchange while minimising the risk of contamination
  • Optional 33 mm threaded caps are available with integrally sealed USP Class VI certified C-Flex® tubing to allow direct sterile transfer of media and cells via pumping or gravity feed
  • Polystyrene construction provides excellent optical clarity and mechanical strength
TC treated surface: Uncharged polystyrene has an uncharged, hydrophobic surface to which cell attachment proteins bind poorly. This results in poor and uneven cell attachment and growth. Tissue culture (TC) treated polystyrene has a negatively charged, hydrophilic surface to which cell attachment proteins bind evenly. This provides a good surface for cell attachment and growth.
Corning® CellBIND® surface: The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields.
More consistent cell attachment resulting in increased cell growth
Adapts cells more quickly to reduced-serum or serum-free conditions
Reduces premature cell detachment from confluent cultures
May eliminate the need for tedious, time-consuming, low stability biological coatings
Requires no refrigeration or special handling and is stable at room temperature
Ultra-Low Attachment surface: The Ultra-Low Attachment surface is a covalently bonded hydrogel surface that is hydrophilic and neutrally charged. It minimises cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert and non-degradable.
Maintains cells in a suspended, unattached state
Prevents stem cells from attachment-mediated differentiation
Prevents anchorage-dependent cells from dividing
Reduces binding of attachment and serum proteins to the substrate
Chambers are sterilized by gamma radiation and certified nonpyrogenic. Manufactured under GMP conditions and 100% leak-tested prior to shipping.
The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields.
 : A variety of optional filling caps are available to allow direct aseptic transfer of media and cells via pumping or gravity feed. Several coupling devices are available on these filling caps with or without integrally sealed USP Class VI certified C-Flex® tubing. Optional filling caps with attached filters with hydrophobic membranes provide for gas exchange and faster aseptic venting during liquid transfers. Extra sterile vented or unvented 33 mm replacement caps are also available. Reusable stacking devices fit between CellSTACK® chambers to keep them level and optimise incubator space while providing clearance for gas exchange.
All caps are 33 mm threaded caps.
  • Recommended working volume:
    625 - 950 ml
  • Seq:
    04
  • Growth area:
    3180 cm²
  • Type:
    5 chambers
  • Cat. No.:
    734-4061
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
  •  
    Description
  • CORN3732
    Universal cap, polyethylene, 33 mm, with vented overcap, double-bagged, sterile
    1 * 4 ST - 46,80 €
  •  
    Description
  • 734-4167
    Solid cap, polyethylene, 33 mm, sterile
    1 * 6 ST - 24,50 €
  •  
    Description
  • 734-4166
    Vent cap, polyethylene, 33 mm, 0,2 µm membrane, sterile
    1 * 6 ST - 51,30 €
  •  
    Description
  • 734-4068
    Fill cap with male MPC; Int. Ø: 9,525 mm
    1 * 4 ST - 293,00 €
  •  
    Description
  • 734-4066
    Fill cap with male MPC; Int. Ø: 6,35 mm
    1 * 4 ST - 316,00 €
  •  
    Description
  • 734-4063
    Fill cap with female MPC coupling; Int. Ø: 9,525 mm
    1 * 4 ST - 327,00 €
  •  
    Description
  • 734-4062
    Fill cap with female MPC; Int. Ø: 6,35 mm
    1 * 4 ST - 316,00 €
  •  
    Description
  • 734-1208
    Two vented over caps and one solid over cap
    1 * 100 ST - 629,00 €
  •  
    Description
  • 734-1209
    Fill cap with male MPC; Int. Ø: 6,35 mm; Tubing length: 69,85 mm
    1 * 4 ST - 340,00 €
  •  
    Description
  • 734-1205
    Vent cap with tubing and 37 mm hydrophobic glass laminate 1,0 μm vent filter; Int. Ø: 6,35 mm; Tubing length: 69,85 mm
    1 * 4 ST - 442,00 €
  •  
    Description
  • 734-4064
    Corning CellSTACK stacking device, ABS, nonsterile
    1 * 5 ST - 246,00 €
  •  
    Description
  • 734-1110
    Fill cap with tubing and barbed fittings; Int. Ø: 9,525 mm; Tubing length: 457,2 mm
    1 * 5 ST - 297,00 €
  •  
    Description
  • 734-1109
    Fill cap with tubing, FLL; Int. Ø: 3,175 mm; Tubing length: 457,2 mm
    1 * 5 ST - 256,00 €
  •  
    Description
  • 734-1108
    Vent cap with tubing and 50 mm hydrophobic PVDF 0,2 µm vent filter; Int. Ø: 9,525 mm; Tubing length: 69,85 mm
    1 * 5 ST - 472,00 €
  •  
    Description
  • CORN11931
    CellSTACK manifold - 4 Luer and 2 MPC connections
    1 * 4 ST - 589,00 €
  •  
    Description
  • CORN11759
    Barbed cap with AseptiQuik® G series; Int. Ø: 9,525 mm; Tubing length: 609,6 mm
    1 * 5 ST - 1 170,00 €
  •  
    Description
  • CORN11760
    CellSTACK 2 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
    1 * 8 ST - 2 290,00 €
  •  
    Description
  • CORN11762
    CellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
    1 * 4 ST - 1 850,00 €
  •  
    Description
  • 734-3409
    CellSTACK screw cap vent accessory with tubing and 50 mm hydrophobic PVDF 0,2 μm vent filter; Int. Ø: 9,525 mm; Tubing length: 76,2 mm
    1 * 5 ST - 549,00 €
  •  
    Description
  • CORN11758
    Barbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 609,6 mm
    1 * 5 ST - 634,00 €
  •  
    Description
  • CORN11761
    CellSTACK 5 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
    1 * 4 ST - 1 530,00 €
  •  
    Description
  • 734-3410
    CellSTACK screw cap fill accessory with tubing; Int. Ø: 9,525 mm; Tubing length: 381 mm
    1 * 5 ST - 360,00 €