Adapters for Huber circulators
Adapter, M30×1,5 male thread to M38×1,5 female thread
Catalog # HUBE6612
Supplier: HUBER
Jump to:
Product Details & Documents
Specifications
- Description:Adapter, M30×1,5 male thread to M38×1,5 female thread
- Seq:34