Orders LinkContactUsLinkComponent
Adapters for Huber circulators
Adapters for Huber circulators
Adapter, M30×1,5 male thread to 1/2" male thread
Catalog # HUBE6393
Supplier:   HUBER
Adapters for Huber circulators
Catalog # HUBE6393
Supplier:  HUBER
Adapters for Huber circulators
Adapters for Huber circulators
Adapter, M30×1,5 male thread to 1/2" male thread
Price
105,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Adapter, M30×1,5 male thread to 1/2" male thread
  • Seq:
    23