Adapters for Huber circulators
Adapter, M16×1 female thread to 1/2" male thread
Catalog # HUBE6360
Supplier: HUBER
Jump to:
Product Details & Documents
Specifications
- Description:Adapter, M16×1 female thread to 1/2" male thread
- Seq:08