Order Entry
Belgium
Orders LinkContactUsLinkComponent
Scoops, EasyScoop

Scoops, EasyScoop

1500 ml, 200 mm
Catalog # BURK5356-1500
Supplier:   BURKLE
Scoops, EasyScoop
Catalog # BURK5356-1500
Supplier:  BURKLE
Scoops, EasyScoop
Scoops, EasyScoop

1500 ml, 200 mm

Price
52,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sampling scoops made of light aluminium, bright-anodised.

  • Ergonomically designed: With centrally arranged inner grip
  • Considerably shorter than conventional scoops: Easier to take samples from narrow containers
Specifications
  • Capacity:
    1500 ml
  • Length:
    200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.