Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multidishes, Nunc™ UpCell™ surface

Multidishes, Nunc™ UpCell™ surface

48-well Multidish, UpCell™, vented, with lid, 1,1 cm², 0,5 ml
Catalog # 734-2378
Multidishes, Nunc™ UpCell™ surface
Catalog # 734-2378
Multidishes, Nunc™ UpCell™ surface
Multidishes, Nunc™ UpCell™ surface

48-well Multidish, UpCell™, vented, with lid, 1,1 cm², 0,5 ml

Price
269,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

The UpCell™ surface is a temperature-responsive cell culture surface that enables harvesting of cells with high viability and intact surface proteins for culture passaging, single-cell analyses and cell transplantation research. The UpCell™ surface enables harvesting of cell sheets and creation of 3D tissue models held together by normal cell junctions and extracellular matrix deposited by the cells.

  • No trypsinisation - cell surface proteins are preserved
  • High cell viability without scraping
  • Releases adherent cells by reduction of temperature of the cell culture
  • Requires minimal hands-on time
Specifications
  • Recommended working volume:
    0,5 ml
  • Description:
    48-well Multidish, UpCell™, vented, with lid
  • Seq:
    04
  • Culture area:
    1,1 cm²
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    NUNC174922
    Log in for availability
    1 * 12 ST - 55,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.