Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multidishes, Nunc™ UpCell™ surface

Multidishes, Nunc™ UpCell™ surface

96-well Multidish, UpCell™, vented, with lid, 0,33 cm², 0,2 ml
Catalog # 734-2377
Multidishes, Nunc™ UpCell™ surface
Catalog # 734-2377
Multidishes, Nunc™ UpCell™ surface
Multidishes, Nunc™ UpCell™ surface

96-well Multidish, UpCell™, vented, with lid, 0,33 cm², 0,2 ml

Price
376,00 €
Log in for availability
1 * 8 ST
Product Details & Documents

The UpCell™ surface is a temperature-responsive cell culture surface that enables harvesting of cells with high viability and intact surface proteins for culture passaging, single-cell analyses and cell transplantation research. The UpCell™ surface enables harvesting of cell sheets and creation of 3D tissue models held together by normal cell junctions and extracellular matrix deposited by the cells.

  • No trypsinisation - cell surface proteins are preserved
  • High cell viability without scraping
  • Releases adherent cells by reduction of temperature of the cell culture
  • Requires minimal hands-on time
Specifications
  • Recommended working volume:
    0,2 ml
  • Description:
    96-well Multidish, UpCell™, vented, with lid
  • Seq:
    05
  • Culture area:
    0,33 cm²
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    Membrane supports for Nunc™ 12-well multidishes with UpCell™ surface
    NUNC174922
    Log in for availability
    1 * 12 ST - 55,10 €