Round canisters
Blue liter scale, 5 L, Natural, S65, with handle and
measuring scale, 167×167×330 mm
measuring scale, 167×167×330 mm
Catalog # SCTV107722
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps.
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 to 4 L, 10, 25, 50 and 60 L for the collection of voluminous solvents
- Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Thread:S65
- Type:with handle and
measuring scale - Capacity:5 L
- Colour:Natural
- Description:Blue liter scale
- Seq:03
- W×D×H:167×167×330 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



