Orders LinkContactUsLinkComponent
VWR®, pH Electrodes
VWR®, pH Electrodes
pH electrode, micro, Non refillable, micro, 0 - 14, 0...60 °C, Epoxy, Mini DIN
Catalog # 662-1814
Supplier:   VWR Collection
VWR®, pH Electrodes
Catalog # 662-1814
Supplier:  VWR Collection
VWR®, pH Electrodes
VWR®, pH Electrodes
pH electrode, micro, Non refillable, micro, 0 - 14, 0...60 °C, Epoxy, Mini DIN
Price
417,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These pH electrodes are available with refillable electrolyte or non refillable.
Documents
Specifications
  • Connection:
    Mini DIN
  • Type:
    Non refillable, micro
  • Description:
    pH electrode, micro
  • Material:
    Epoxy
  • Temp.:
    0...60 °C
  • pH:
    0 - 14