KIT METAL 2,8MM REINFORCED 2 ML TUBES ,ready-to-use kits with the Precellys24,includes 50 microtubes, prefilled with 2,8mm stainl ess steel beads in 2mL reinforced tubes.Custom design reinforced tubes for bead beating application : o-ring and reinfo 1 * 1 KIT
Catalog # BERT03961-1-008
Supplier: Bertin Instruments
Frequently Bought Together