Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Bevelled, non-graduated, 1 - 200 µl, -, 50 mm, Yellow, 96 tips/rack; 10 racks/case
Catalog # ALPCLW6360
Supplier:   Alpha Laboratories
Pipette tips
Catalog # ALPCLW6360
Supplier:  Alpha Laboratories
Pipette tips
Pipette tips
Bevelled, non-graduated, 1 - 200 µl, -, 50 mm, Yellow, 96 tips/rack; 10 racks/case
Price
145,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
  • Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
  • Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
  • With graduations to visually check volume
  • Suitable for all major pipettor brands
Specifications
  • Colour code:
    Yellow
  • Packed:
    96 tips/rack; 10 racks/case
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Length:
    50 mm
  • Seq:
    04
  • Tip style:
    Bevelled, non-graduated