Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Bevelled, non-graduated, 200 - 1000 µl, -, 76 mm, Blue, Bulk, 1000 tips/bag
Catalog # ALPCLW6471
Supplier:   Alpha Laboratories
Pipette tips
Catalog # ALPCLW6471
Supplier:  Alpha Laboratories
Pipette tips
Pipette tips
Bevelled, non-graduated, 200 - 1000 µl, -, 76 mm, Blue, Bulk, 1000 tips/bag
Price
90,60 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
  • Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
  • Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
  • With graduations to visually check volume
  • Suitable for all major pipettor brands
Specifications
  • Colour code:
    Blue
  • Packed:
    Bulk, 1000 tips/bag
  • Sterile:
    -
  • Volume:
    200 - 1000 µl
  • Length:
    76 mm
  • Seq:
    10
  • Tip style:
    Bevelled, non-graduated