Pipette tips
Micro, ultrafine, 0,1 - 10 µl, -, 32 mm, Natural, Bulk, 1000 tips/bag
Catalog # ALPCLW1154
Supplier: Alpha Laboratories
Jump to:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specifications
- Colour code:Natural
- Packed:Bulk, 1000 tips/bag
- Sterile:-
- Volume:0,1 - 10 µl
- Length:32 mm
- Seq:03
- Tip style:Micro, ultrafine
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



