METHACRYLIC ACID-ETHYLACRYLATE COPOLYMER 1 * 1 items
Catalog # EDQMM0610030
Supplier: EDQM
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Pralidoxime Chloride, Reference Standard
Supplier: USP
Buffer Solution pH 4.006 1 * 1 L
Supplier: PanReac AppliChem
PIROCTONE OLAMINE 1 * 500 mg
Supplier: Merck
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Milrinone, Reference Standards
Supplier: USP
VWR®, Certified Reference Material, Cleveland Open Cup flash point
Supplier: VWR Chemicals
Ferric ammonium sulphate solution R5 Reag. Ph. Eur. 1037704
Supplier: VWR Chemicals
Supelco® Organic Reference Standard, Myristoleic Acid
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
ACQUITY ARC RI START-UP SOLUTION 1 * 1 KIT
Supplier: WATERS