Orders LinkContactUsLinkComponent
Objectives
EC-Planachromatic Neofluar, M27, N.A. of 0,3, WD of 5,2 mm
Catalog # ZEIS4203409901000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4203409901000
Supplier:  ZEISS
Objectives
Objectives
EC-Planachromatic Neofluar, M27, N.A. of 0,3, WD of 5,2 mm
Price
1 106,96 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    10×
  • Description:
    EC-Planachromatic Neofluar, M27, N.A. of 0,3, WD of 5,2 mm