Objectives
EC-Planachromatic Neofluar, M27, N.A. of 0,16, WD of 18,5 mm
Catalog # ZEIS4203309901000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Magnification:5×
- Description:EC-Planachromatic Neofluar, M27, N.A. of 0,16, WD of 18,5 mm
- Seq:16