Orders LinkContactUsLinkComponent
Percoll®, Siam
Percoll®, Siam
Percoll®
Catalog # P1644-100ML
Percoll®, Siam
Catalog # P1644-100ML
Percoll®, Siam
Percoll®
Price
198,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
  • Non toxic
  • Ideal for use with biological materials
Documents
Specifications
  • Description:
    Percoll®
  • Seq:
    03