Automatic burettes, Pellet pattern, class AS
50 ml, 0,10 ml
Catalog # BIBB3296/06AS
Supplier: SciLabware
Jump to:
Product Details & Documents
Burettes made of chemically resistant PYREX® borosilicate glass.
- Automatic zero facility
- Simple operation
- White enamel graduations
- 1,5 mm bore PTFE key stopcock
: Complies with ISO 385 and DIN 12700.
Specifications
- Capacity:50 ml
- Seq:03
- Division:0,10 ml
Accessories
Product Information
- Catalog #AvailabilityPrice
- Blowing ball, rubber, for automatic burettes, Pellet patternBlowing ball, rubber, for automatic burettes, Pellet patternBIBB3297/14ASLog in for availability1 * 1 ST - 32,20 €
Product Information
- Catalog #AvailabilityPrice
- Reservoir bottle, glass, clear, 2000 ml, with 29/32 jointReservoir bottle, glass, clear, 2000 ml, with 29/32 jointBIBB3297/12Log in for availability1 * 1 ST - 66,30 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



