Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Forceps, precision

Straight, short fine tips, 4, 110 mm
Catalog # 232-0216
Supplier:   OUTILS RUBIS
Forceps, precision
Catalog # 232-0216
Supplier:  OUTILS RUBIS
Forceps, precision
Forceps, precision

Straight, short fine tips, 4, 110 mm

Price
30,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Forceps with very precisely machined tips. Ideal for applications that require a high degree of precision, including microscopy, anatomy, biology, microbiology, gene technology and electron microscopy.

  • Acid-resistant
  • Resistant against hydrogen fluoride and nitric acid
  • 100% anti-magnetic
Specifications
  • Type:
    4
  • Description:
    Straight, short fine tips
  • Length:
    110 mm