Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sampling siphons

Sampling siphons

Sampling siphon, PE, 75 ml, 500 mm
Catalog # BURK5387-0050
Supplier:   BURKLE
Sampling siphons
Catalog # BURK5387-0050
Supplier:  BURKLE
Sampling siphons
Sampling siphons

Sampling siphon, PE, 75 ml, 500 mm

Price
16,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Disposable plunging samplers acting like siphons.

  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler

If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.

Specifications
  • Capacity:
    75 ml
  • Description:
    Sampling siphon
  • Length:
    500 mm
  • Material:
    PE
  • Seq:
    01