Orders LinkContactUsLinkComponent
Stopcocks, with GL screw thread
8 mm, GL 25
  229-0059
 :   Bohlender
Stopcocks, with GL screw thread
  229-0059
 :  Bohlender
Stopcocks, with GL screw thread
Stopcocks, with GL screw thread
8 mm, GL 25
203,00 €
1 * 1 ST
PTFE.
  • Excellent chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −30 to +150 °C
  • Pressure resistant up to a maximum of 6 bar, vacuum suitable
Two-way stopcocks with straight bore and two threaded connectors or three-way stopcocks with L-shaped or T-shaped bore with three threaded connectors. Cylindrical stopcock for good leakproof properties, stop valve with flow direction marking. To distribute liquids or gases. Fast and simple interruption of the flow. Connection of hoses or tubes using GL laboratory screw connections.
  • Hole Ø:
    8 mm
  • Thread:
    GL 25