Orders LinkContactUsLinkComponent
Closing cones, Combi-Stoppers
Closing cones, Combi-Stoppers
Closing cones for needle-free injection sites and seal pre-filled syringes, White
Catalog # BRAU4495209
Supplier:   B. Braun
Closing cones, Combi-Stoppers
Catalog # BRAU4495209
Supplier:  B. Braun
Closing cones, Combi-Stoppers
Closing cones, Combi-Stoppers
Closing cones for needle-free injection sites and seal pre-filled syringes, White
Price
53,80 €
Log in for availability
1 * 100 ST
Product Details & Documents
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
  • Unique multifunctional design
  • Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.
Specifications
  • Colour:
    White
  • Description:
    Closing cones for needle-free injection sites and seal pre-filled syringes
  • Seq:
    03