Pipette tips
Gilson type, 100 - 1000 µl, -, Blue, Gilson P500/P1000, Eppendorf, Socorex, Nichiryo, HTL, Bulk (10×1000)
Supplier: DELTALAB
Jump to:
Product Details & Documents
Specifications
- Packed:Bulk (10×1000)
- Sterile:-
- Volume:100 - 1000 µl
- Seq:16
- Tip style:Gilson type
- Tip colour:Blue
- For use with:Gilson P500/P1000, Eppendorf, Socorex, Nichiryo, HTL