Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-channel pipettes, mechanical, variable volume, SoftGrip™

30 - 300 µl, 0,2, ±1,0 - ±2,4 %, ±0,5 - ±1,2 %, Red, 300 µl
  HAMI55019-56
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
  HAMI55019-56
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
Multi-channel pipettes, mechanical, variable volume, SoftGrip™

30 - 300 µl, 0,2, ±1,0 - ±2,4 %, ±0,5 - ±1,2 %, Red, 300 µl

 
998,00 €
   
1 * 1 ST
 

Hamilton SoftGrip pipettes are available as 8-channel and 12-channel adjustable volume manual pipettes in two volume ranges covering 5 to 50 µl and 30 to 300 µl.

  • Long tip ejector lever and movable lower body can be rotated a full 360º relative to the handle to provide maximum comfort for left- or right-handed use
  • Colour-coded plunger buttons and labels for quick volume identification
  • Easily activated tip ejector lever releases all the tips in single stroke
  • Can be autoclaved without the need for disassembly or recalibration

Please note: Accuracy and precision is guaranteed when used with Hamilton Universal AdvanTip™ precision pipette tips.

 
  • Colour code:
    Red
  • Volume:
    30 - 300 µl
  • Increment:
    0,2
  • Accuracy:
    ±1,0 - ±2,4 %
  • Imprecision:
    ±0,5 - ±1,2 %
  • Recommended tips:
    300 µl
Accessories

 

  •  
    Description
     
     
  • HAMI1987-01
    Calibration labels for pipettes
       
    1 * 10 ST - 34,70 €

 

  •  
    Description
     
     
  • HAMI34002
    Pipette calibration key, for simple calibration in the field
       
    1 * 1 ST - 9,80 €

 

  •  
    Description
     
     
  • HAMI54756-01
    Ejector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µl
       
    1 * 1 ST - 23,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.