Multi-channel pipettes, mechanical, variable volume, SoftGrip™
5 - 50 µl, 0,02, ±0,8 - ±2,0 %, ±0,35 - ±2,0 %, Brown, 200 µl
N° de catalogue HAMI55019-50
Fournisseur: HAMILTON BONADUZ
Multi-channel pipettes, mechanical, variable volume, SoftGrip™
N° de catalogue HAMI55019-50
Fournisseur: HAMILTON BONADUZ
Jump to:
Product Details & Documents
Hamilton SoftGrip pipettes are available as 8-channel and 12-channel adjustable volume manual pipettes in two volume ranges covering 5 to 50 µl and 30 to 300 µl.
- Long tip ejector lever and movable lower body can be rotated a full 360º relative to the handle to provide maximum comfort for left- or right-handed use
- Colour-coded plunger buttons and labels for quick volume identification
- Easily activated tip ejector lever releases all the tips in single stroke
- Can be autoclaved without the need for disassembly or recalibration
Please note: Accuracy and precision is guaranteed when used with Hamilton Universal AdvanTip™ precision pipette tips.
Specifications
- Colour code:Brown
- Volume:5 - 50 µl
- Increment:0,02
- Accuracy:±0,8 - ±2,0 %
- Imprecision:±0,35 - ±2,0 %
- Recommended tips:200 µl
Accessories
Product Information
- N° de catalogueDisponibilitéPrix
- Calibration labels for pipettes
Product Information
- N° de catalogueDisponibilitéPrix
- Pipette calibration key, for simple calibration in the fieldPipette calibration key, for simple calibration in the fieldHAMI34002Log in for availability1 * 1 ST - 9,80 €
Product Information
- N° de catalogueDisponibilitéPrix
- Ejector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlEjector sleeve for SoftGrip™ single channel pipette, 500 - 1000 µlHAMI54756-01Log in for availability1 * 1 ST - 23,80 €