Orders LinkContactUsLinkComponent
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit), 30 m
Catalog # YSIA6051030-30
Supplier:   YSI ENVIRONMENTAL
Cables for Multi-Parameter Meter Pro 1030, Handheld
Catalog # YSIA6051030-30
Supplier:  YSI ENVIRONMENTAL
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cables for Multi-Parameter Meter Pro 1030, Handheld
Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit), 30 m
Price
1 650,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cable with built-in temperature and conductivity and with a port for a pH or ORP sensor (incl. Cable Management Kit)
  • Length:
    30 m
  • Seq:
    05