Order Entry
Belgium
Orders LinkContactUsLinkComponent
Selenium, powder

Selenium, powder

Glass bottle for solids, 50 g
Catalog # A12592.18
Toxic
Health
Selenium, powder
Catalog # A12592.18
CAS Number:  7782-49-2
Selenium, powder
Selenium, powder

Glass bottle for solids, 50 g

CAS Number:  7782-49-2
Price
46,60 €
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • UN:3283
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.