Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Selenium, powder

Glass bottle for solids, 1.000 g
Catalog # A12592.0B
Toxic
Health
Selenium, powder
Catalog # A12592.0B
CAS Number:  7782-49-2
Selenium, powder
Selenium, powder

Glass bottle for solids, 1.000 g

CAS Number:  7782-49-2
Price
509,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • UN:3283
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications