Order Entry
Belgium
Orders LinkContactUsLinkComponent
Syringes for CTC analytics systems

Syringes for CTC analytics systems

Removable needle, 22, 1000 µl, 51 mm, 0,72 mm, LC, 1MR-CTC-GT-LC(0.4) *
Catalog # 554-1465
Syringes for CTC analytics systems
Catalog # 554-1465
Syringes for CTC analytics systems
Syringes for CTC analytics systems

Removable needle, 22, 1000 µl, 51 mm, 0,72 mm, LC, 1MR-CTC-GT-LC(0.4) *

Price
113,00 €
Log in for availability
1 * 1 ST
Specifications
  • Ext.Ø:
    0,72 mm
  • Gauge:
    22
  • Type:
    1MR-CTC-GT-LC(0.4) *
  • Type of tip:
    LC
  • Volume:
    1000 µl
  • Description:
    Removable needle
  • Length:
    51 mm
  • Seq:
    10
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.